Wound Clip, EZ
Katalog # STOE59027
Leverantör: STOELTING
Specifikationer
- Beskrivning:EZ Wound Clip™ Clips
- Storlek:9 mm
- Förp.:1.000
Specifikationer
Om denna produkt
This EZ Clip™ wound closing kit is perfect for rapidly closing wounds, providing maximum holding force with minimal trauma.
- Easier and faster to use than conventional sutures and needles
- After healing, the clips can be easily removed with the clip remover
- Pyrogen-free and nonsterile
Delivery: EZ Clip™ Kit includes the Applier, 9 mm wound clips (case of 1000) and the remover. Individual components may be ordered separately.
Packaging: Racks of 9 mm clips are individually packaged in heat sealed, peelaway envelopes for cleanliness and ease of handling and storage. EZ Clips™ are packaged 20 on a rack, with 10 racks per box (200 total), or 50 racks per case (1000 total).