Hightextra® Smooth Wipes
Katalog # ECOLHTS0060R05300W
Leverantör: ECOLINE NV SA
Specifikationer
- Färg:Vit
- Beskrivning:Smooth wipes, 68 gsm
- Förp.:5 Roll
Specifikationer
Om denna produkt
The combination of woodpulp and continuous polyester fibres creates a super strong wipe, even when used wet, that copes with tougher cleaning too.