ContactUsLinkComponent
DSDM® Dehydrated Culture Media
DSDM® Dehydrated Culture Media
Catalog # DOEH971231244
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
DSDM® Dehydrated Culture Media
Catalog # DOEH971231244
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Förp.:
    1
  • Beskrivning:
    DSDM® dehydrated culture media, 200 g
Specifications
About this item
Ready-to-prepare dehydrated media for the detection and enumeration of diastatic wild yeast population in brewing culture yeast and beer samples.
  • Powder provides highest flexibility
  • Easier interpretation
  • Incorporated colour indicator
  • High selectivity
  • Functionality checked by strains from practice
  • Effortless an-/ aerobic incubation
DSDM® convinces with highest selectivity and specificity, also for detection in culture yeast. It is mainly used as a liquid medium, but also by adding agar-agar as a solid medium. By selecting appropriate components, only the growth of super-attenuating yeast is promoted, while culture yeast is inhibited. An additional colour indicator helps to interpret the results by showing a colour change.