Cerex® OFXQ 96 Well Plate, RCH
Catalog # TCAT635-0101RCH
Leverantör: Tecan
Specifikationer
- Beskrivning:Plate RCH Cerex OFXQ
- Phase:OFXQ
- Förp.:1
Specifikationer
Om denna produkt
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
- Ion-exchange
- Liquid handler compatible
A proprietary polymer resin blend of ion exchange chemistry providing unique selectivity. Especially designed for scientists working with oral fluid matrixes.