ContactUsLinkComponent
illustra™ plasmidPrep Midi Flow Kits
illustra™ plasmidPrep Midi Flow Kits
Catalog # 28-9042-66
Supplier:  Cytiva
CAS Number:  
undefined
illustra™ plasmidPrep Midi Flow Kits
Catalog # 28-9042-66
Supplier:  Cytiva
CAS Number:  
Specifications
  • Förp.:
    5 Tests
  • Method / Format:
    Column (anion exchange)
  • Prep size:
    Midi
  • Refine target molecule:
    Plasmid
    Larger vectors (Cosmid, BAC, PAC, Fosmid)
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Beskrivning:
    illustra™ plasmidPrep midi flow kit, 5 preps
  • Seq:
    01
  • Provstorlek:
    25 - 150 ml
  • Benämning:
    illustra™ plasmidPrep midi flow kit
Specifications
About this item
The illustra™ plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume.
  • Provides plasmid DNA with excellent purity and low endotoxin levels
  • Increased ease of use due to minimal pipetting volume changes, colour-coded kit components, and pre-equilibrated columns
  • Purification is in single-use format, gravity Fast-Flow columns
The illustra™ plasmidPrep Midi Flow Kit can be used to process bacterial sample volumes of 25 to 50 ml for high copy number plasmid DNA and up to 500 ml for low copy number plasmid DNA. The kit consists of a high-performance, anion exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA.
The illustra™ plasmidPrep Midi Flow Kit is ideal for the rapid extraction and purification of high quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification.
Delivery: Kits include Fast-Flow plasmid columns, resuspension, lysis, neutralisation, wash, elution, and TE buffers, and a full protocol booklet with a detachable, quick reference protocol card.