Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Saltsyra
Saltsyra 1 mol/l (1 N)
Saltsyra 1 mol/l (1 N)
FFCHFF2275000
:
FF CHEMICALS
:
Saltsyra 1 mol/l (1 N)
FFCHFF2275000
:
FF CHEMICALS
:
Förp.:
5 L
Seq:
5190
:
:
8,II
:
36.46 g/mol
:
231-595-7
:
1789
:
GHS05
:
Omgivande
:
7647-01-0
:
HCl
:
MFCD00011324