Refractix™ Universal Digital Refractometer
Digital refractometer with color touch screen
Katalog # REIC13910000
Leverantör: REICHERT
Gå till:
Product Details & Documents
Refractometer provides laboratory performance in a compact portable platform.
- Sapphire prism
- 250 measurement scale capacity with 50 scales pre-loaded
- 2,8" color touch screen and physical buttons
- High resolution 1500 CCD linear image sensor with benchtop accuracy
- Measurement log stores last 5000 measurements
- Bluetooth and USB-C connectivity to RefractixSync PC software
- Rechargeable battery with AC power option
- Designed, engineered, and manufactured in the USA
The Refractix™ refractometer is innovative, thoughtfully designed, portable, and defies classification. Can be used in the laboratory with benchtop accuracy/precision and can be used as a portable on the go. 250 measurement scale capacity with 50 scales preloaded and found in scale groups for food/beverage, chemical, beer/wine/spirits, automotive, veterinary and favorites.
View real time measurements and download 5000 measurement log through RefractixSync software PC based App. Laboratory performance in a value added and affordable platform.
Specifikationer
- Beskrivning:Digital refractometer with color touch screen
- Cat. No.:REIC13910000
- Noggrannhet:0,0001 nD (0,1/100 g brix) [precision]
- Område för temperaturkompensation:10...40 °C (50...104 °F)
- W×D×H:17,6×7,3×4,2 cm
- Mätområde:1,3300 - 1,5600 nD (0,0 - 85,0/100 g Brix ICUMSA)
- Vågar:250 scales
- Strömförsörjning:Rechargeable Li-ion battery and AC outlet power option