Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethyleneimine
Polyethyleneimine, M.W. ∼10,000 (by GPC), branched
Polyethyleneimine, M.W. ∼10,000 (by GPC), branched
Polyethyleneimine, M.W. ∼10,000 (by GPC), branched
Supplier:
Spectrum Chemical
Certificates
Expand image
Polyethyleneimine, M.W. ∼10,000 (by GPC), branched
Supplier:
Spectrum Chemical
About this item
CAS Number:
9002-98-6
:
Formula:
(CH2CH2NH)n
MDL Number:
MFCD00084427
Density:
1,030
:
GHS07
Storage Temperature:
Omgivande