Valved Dolomite Fold Flat Respirators, EAGLE FFP2
Catalog # PRTWP291WHR
Supplier: Portwest
Specifications
- Beskrivning:EAGLE FFP2 valved Dolomite flat-fold respirator mask
- Förp.:10
Specifications
About this item
Innovative compact folding mask with seven dynamic parts to provide excellent facial movement with maximum comfort.
- Seven ergonomic points which adapt to different face shapes
- High performance exhalation valve provides comfort in hot and humid conditions
- Flexible shape allows the mask to be folded flat
- Individually wrapped inside hygienic polybag
- Adjustable nose bar for improved fit and reduced fogging of glasses
- Foam padding around the nose bridge for additional comfort
Valved and with inner foam for increased comfort for the wearer. Dolomite optional clogging test (D) to provide more comfortable breathing and longer lasting filtration performance. An individual polybag and special flat shape make it easy to store and vending machine ready.
Certifications: CE-CAT III.