USB Rechargeable LED Head Light
Catalog # PRTWPA70BKR
Supplier: Portwest
Specifications
- Beskrivning:USB rechargeable LED head light
- Förp.:1
Specifications
About this item
This USB rechargeable head light features multiple light settings, tilt control, and a soft adjustable strap making it comfortable to wear.
- Brightness: 150 lumens
- Run time: 4 h
- Recharge time: 5 h
- Micro USB charging cable included
- Tilt control for different working positions
- Impact- and water-resistant
- Retail box aids presentation for retail sales
This light is rated to IP X4, making it wind- and water-resistant, and perfect for all weather conditions.