Lens Cleaning Wipes
Catalog # PRTWPA01WHR
Supplier: Portwest
Specifications
- Färg:Vit
- Beskrivning:Lens cleaning wipes
- Förp.:100
Specifications
About this item
Lens Cleaning Wipes are packaged in a handy dispenser box which is pre-assembled for immediate use.
- 100 towelettes per box
- Low-linting construction for minimal contamination
- Each towelette is individually wrapped
- Suitable for all Portwest safety eyewear
- Anti-static
- No alcohol content
Silicone free wipes are ideal for manufacturing, paint applications, electronics, and glass handling applications where silicone is problematic.
Dispenser box measures 130×200 mm, and holds 100 individually packaged lens cleaning wipes.