Salpetersyra concentrate for dilution to 1L, 0.1 M HNO3 (0.1N) volumetrisk lösning, Fluka™
Catalog # 32042-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Förpackningstyp:Ampoule
- Förp.:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:63.01 g/mol
Boiling Pt:100 °C
Density:1,065 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349