ContactUsLinkComponent
Propidium iodide
Propidium iodide
Catalog # CAYM10009957-1
Supplier:  Cayman Chemical
CAS Number:  
Propidium iodide
Catalog # CAYM10009957-1
Supplier:  Cayman Chemical
CAS Number:  
Specifications
  • Förp.:
    1
  • Seq:
    0001
Specifications
About this item
 :
 :668.4 g/mol
EINECS:247-081-0
 :GHS07
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921