Human thrombin (from plasma)
Katalog # 605195-5000
Leverantör: Merck Millipore (Calbiochem)
CAS-nummer:
Specifikationer
- Enzymnamn:Thrombin
- Enzymaktivitet:≥2800 NIH units/mg
- Art.nr:605195-5000
Specifikationer
Om denna produkt
Designed for use in thrombin clotting time tests. Key enzyme that activates factor XIII and converts fibrinogen into fibrin.