Etylmetylketon
Catalog # ABSO1091
Supplier: ABSOR
CAS Number:
Specifications
- Förp.:5 L
- Seq:5190
Specifications
About this item
:
Merck Index:13,06097
Boiling Pt:79,6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0,8054 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Omgivande
EINECS:201-159-0
UN:1193
ADR:3,II
:72.11 g/mol
REACH:01-2119457290-43
:GHS02
:GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648