Petroleumeter, 80…110 °C
Supplier: ABSOR
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:80…110 °C (1013 hPa)
Storage Temperature:Omgivande
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09