Reprospher 100 PEI HPLC Columns
Supplier: DR MAISCH
Certificates
About this item
Reprospher media are based on a 100 Å ultrapure silica and are fully scalable from UPLC (1,9 µm particles) to preparative and process scale applications.
Specifications
- Catalog No:
- MAISRS10.PEI.S2546
- MAISRS15.PEI.S1520
- MAISRS15.PEI.S2530
- MAISRS13.PEI.S0330
- MAISRS15.PEI.S1503
- MAISRS13.PEI.S1546
- pH område:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g