Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Elektroder
pH-elektroder
Accessories for Waterproof pH electrode GE 114-WD, GHM Messtechnik GmbH
Accessories for Waterproof pH electrode GE 114-WD, GHM Messtechnik GmbH
Accessories for Waterproof pH electrode GE 114-WD, GHM Messtechnik GmbH
Leverantör:
Greisinger
Certifikat
Expand image
Accessories for Waterproof pH electrode GE 114-WD, GHM Messtechnik GmbH
Leverantör:
Greisinger
Om denna produkt