
FURA-2 pentapotassium salt ≥95%
Catalog # AATB21025
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 mg
- Seq:2070
Specifications
About this item
:
:832 g/mol
ADR:5,II
:GHS07
CAS Number:113694-64-7
Formula:C₂₉H₂₂K₅N₃O₁₄