Helixyte™ Green Nucleic Acid Gel Stain 10,000X DMSO Solution
Supplier: AAT BIOQUEST
Certificates
About this item
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
- Same spectral properties to SYBR® green
- High sensitivity
- Useful for nucleic acid labeling in gels
Caution: For research use only.
Specifications
- Renhet:95%