Order Entry
Sweden
ContactUsLinkComponent
Nuclear Green™ LCS1
Nuclear Green™ LCS1
Katalog # AATB17540
Leverantör:  AAT BIOQUEST
CAS-nummer:  
undefined
Nuclear Green™ LCS1
Katalog # AATB17540
Leverantör:  AAT BIOQUEST
CAS-nummer:  

Specifikationer

  • Förp.:
    0,5 mL
  • Beskrivning:
    Nuclear Green™ LCS1
  • Renhet:
    90%

Specifikationer

Om denna produkt

Our Nuclear Green™ LCS1 is a fluorogenic, DNA-selective and cell-permeant dye for analysing DNA content in living cells.

  • Cell-Permeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence

5 mM DMSO solution.

Caution: For research use only.