PSA™ iFluor® 555 Imaging Kit with Goat Anti-Rabbit IgG
Katalog # AATB45220
Leverantör: AAT BIOQUEST
CAS-nummer:
Specifikationer
- Förp.:100 Tests
- Beskrivning:Can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 - 50 times higher than the widely used tyramide (TSA) reagents
Specifikationer
Om denna produkt
iFluor® 555 PSA kit is a much superior replacement for Alexa Fluor 555 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
- Higher efficiency
- Improved fluorescence signal
- Less consumption of primary antibody
- 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Packaging: Plastic vial.
Caution: For research use only.