ReadiLink™ Biotin Oligo and ssDNA Labelling Kit
Leverantör: AAT BIOQUEST
Certifikat
Om denna produkt
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Efficient labelling
- Contains all essential reagents
- Multiple applications
The resulting biotinylated oligo and ssDNA probes are ideally suited for a variety of biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridisations..
Packaging: Packaged in plastic vials.
Caution: For research use only.