Transfectamine™ 5000 Transfection Reagent
Transfectamine™ 5000 Transfection Reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells.
- Easy-to-use, minimal steps required
- High transfection efficiency
- Works with a wide variety of eukaryotic cells
- Effective on dissociated cells and slices
: For research use only.
- Bioproduktion:Ja
- Cellspecifik:Mammalian
- DNA och siRNA:Ja
- DNA:Ja
- shRNA:Ja
- Förvaringstemperatur:Freeze (< –15 °C); minimise light exposure