FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit)
Catalog # AATB20001
Supplier: AAT Bioquest
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:FluoroQuest™ anti-fading kit I *optimized for slide imaging*
Specifications
About this item
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
- Preserves fluorescence
- Prevents photobleaching
- Ready to use
- Contains all necessary components
Caution: For research use only.