ReadiLink™ Rapid mFluor™ Blue 570 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
AATB1120
:
- Förp.:2 Tests
- Beskrivning:ReadiLink™ rapid mFluor™ blue 570 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Blue-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
: For research use only.