ReadiLink™ xtra Rapid XFD594 Antibody Labelling Kit *BSA-Compatible, XFD594 Same Structure to Alexa Fluor™ 594* (2 Labellings)
AATB1982
:
- Förp.:2 Tests
- Beskrivning:ReadiLink™ xtra rapid XFD594 antibody labeling kit 'BSA-compatible, XFD594 same structure to Alexa Fluor™ 594'
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
: For research use only.