5-Bromo-2'-deoxyuridine (broxuridine) ≥95%
Catalog # AATB17030
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:25 mg
- Seq:2315
Specifications
About this item
:
:307.10 g/mol
ADR:5,II
:GHS07
CAS Number:59-14-3
Formula:C₉H₁₁BrN₂O₅
MDL Number:MFCD00006529