Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Media
Dehydrerade mikrobiologimedia
VWR® Modified Tryptone Soya Broth with Novobiocin
VWR® Modified Tryptone Soya Broth with Novobiocin
VWR® Modified Tryptone Soya Broth with Novobiocin
Catalog # 411120AF
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
VWR® Modified Tryptone Soya Broth with Novobiocin
Catalog # 411120AF
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Förp.:
10
Beskrivning:
Modified tryptone soya broth with novobiocin
Specifications
About this item
Product Details
Documentation
Product Details
Documentation