Drop-Out Mix Complete without Yeast Nitrogen Base
Katalog # USBID9515-100G
Leverantör: US Biological
CAS-nummer:
Specifikationer
- Förp.:100 g
- Beskrivning:Drop-out mix complete without yeast nitrogen base (DO, dropout, powder)
Specifikationer
Om denna produkt
Drop-out mixes are used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
- Convenient powders
- Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.