Chimney with Fan for Muffle Furnaces
Catalog # NABE6000140311
Supplier: NABERTHERM
Specifications
- Beskrivning:Chimney with fan for Nabertherm muffle furnaces
- Förp.:1
Specifications
About this item
Using chimney with fan exhaust gases are better removed from the furnace and discharged.
The B500 - P580 controllers can be used to switch the chimney with fan automatically (not for models L(T) 15.., L 1/12, LE 1/11, LE 2/11).