ContactUsLinkComponent
Gas cartridge, CG1750
Gas cartridge, CG1750
Katalog # CAMP2182099
Leverantör:  CAMPING GAZ
undefined
Gas cartridge, CG1750
Katalog # CAMP2182099
Leverantör:  CAMPING GAZ
Specifikationer
  • Beskrivning:
    Gas cartridge, CG 1750, 175 g capacity, weight: 280 g, Ø×H: 68×162 mm
  • Förp.:
    12
Specifikationer
Om denna produkt
Campingaz® gas cartridge CG1750 with safety valve.
  • Filled with 70% butane and 30% propane gas
  • Equipped with high security self-sealing valve
  • Can be disconnected and reconnected even when the cartridge is not empty
  • Ideal for sporadic, moderate use