3M™ Combination Filter
Catalog # MMMA7100260645
Supplier: 3M
Specifications
- Beskrivning:Combination filter
- för användning med:3M™ full facepiece reusable respirator FF-300 series, 3M™ full facepiece reusable respirator FF-600 Series, 3M™ powered air respirator PF-600E
- Förp.:1
Specifications
About this item
For effective protection against a range of particulates, gases and vapours or combination hazards, use 3M™ Filters DT-Series. Suitable for use with certain 3M™ half and full-face tight-fitting respirators in negative-pressure mode as well as 3M™ Powered Air Respirator System PF-600E+ Series.
- Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
- Features a DIN40 mm threaded connection
- Please note DT-Series filters were previously known as Pro2000 Filters
- Suitable for use with 3M™ Powered Air Respirators PF-602E+ and PF-602E-ASB. Note: A1 classification when used with powered air systems
Protection type: Acid gas, ammonia, iorganic vapor, mercury, multi-gas, organic vapour, particulate
Recommended application: Demolition, grinding, metalworking