Proteinfingeravtrycksbestämning
Catalog # BE-408
Supplier: G-Biosciences
Specifications
- Beskrivning:Proteinfingeravtrycksbestämning
- Förp.:1 KIT
Specifications
About this item
In this lab activity, students learn to perform simple protein isolation procedures to isolate the protein fingerprint from various fresh tissues. They will compare the protein fingerprints of four different tissues to understand that the function of a particular organ is due to the proteins that are localised to the specific organ. Also included in this kit are four dried protein samples (from mouse liver, brain, heart and lungs) to compare as a control, if fresh tissues cannot be obtained.
- Hantering av proteinprover
- Proteinmolekyler som livets byggstenar
- Proteinelektrofores för som analysmeteod
- Proteindiversitet i djurriket
Behövs också: Proteinelektroforesutrustning.
Delivery: Levereras med de beståndsdelar som behövs för praktisk experimentering för 6 labgrupper. Levereras med lärarhandbok och separat elevhandbok (handböcker på engelska).