Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Belysning
Halogenlampor
Halogen-spotlight, HALOSPOT® 48, med aluminiumreflektor
Halogen-spotlight, HALOSPOT® 48, med aluminiumreflektor
Halogen-spotlight, HALOSPOT® 48, med aluminiumreflektor
Catalog # 139-0144
Supplier:
OSRAM
Expand image
Halogen-spotlight, HALOSPOT® 48, med aluminiumreflektor
Catalog # 139-0144
Supplier:
OSRAM
Specifications
Botten:
GY4
Förp.:
1
Effekt (W):
20 W
Spänning:
12 V
Specifications
About this item
Product Details
Product Details