Nukleinsyrakvantifiering
Katalog # BE-318
Leverantör: G-Biosciences
Specifikationer
- Beskrivning:Nukleinsyrakvantifiering
- Förp.:1 KIT
Specifikationer
Om denna produkt
The kit utilises the principle of diffusion of nucleic acids on a nylon membrane to determine their concentration. No spectrophotometers required.
- Ingen dyr utrustning behövs
- Snabb kvantifiering DNA- och RNA-prover
- Tillräckligt med reagenser för mer än 50 analyser
Behövs också: DNA-prover.
Nucleic acid concentration can be measured by measuring the diameter of diffused nucleic acid spots or comparing the colour density of the spots with a set of known standards. Accurate and fast DNA and RNA concentrations can be determined with as little as 1 μl of DNA sample.
Delivery: Levereras med 50 analysremsor förutom alla buffertar och skalor som krävs för DNA- eller RNA-kvantifiering.