Dataloggenheter, USBLink
Catalog # 620-1795
Leverantör: FOURIER SYSTEMS
Specifikationer
- Gränssnitt:USB
- Beskrivning:Datalogger, USBLink
- Förp.:1
Specifikationer
Om denna produkt
The USBLink is a sensor interface, converting any computer into a science data logging learning station. The USBLink simple 'plug-and-play' unit is highly functional, featuring four-input interface, with an option for up to eight simultaneously connected sensors via USB to the computer. The unit offers automatic sensor recognition, fast sampling rate and sample streaming of up to 10,000 samples per second.
- Lätta att hantera
- Simultan datainsamling för upp till 8 sensorer samtidigt
- Komplett utbud av Fourier-sensorer finns tillgängligt
- Ansluts direkt via USB-port
- Levereras med programvaran MultiLab