Scotch-Brite™ 1000 Diamond Floor Brush
Catalog # MMMA7100262369
Supplier: 3M
Specifications
- Ø:431,8 mm
- För:Floor
- Beskrivning:Floor brush, round, 1000 grit, 175 - 600 min⁻¹ machine speed
- Förp.:2
Specifications
About this item
Scotch-Brite™ Diamond floor brush 1000 series is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step. Brush 1000 is a coarser grit brush intended for daily use on floors where low gloss is desired.
- Provides both cleaning and gloss recovery in one step
- Conformable, flexible diamond bristles reach low spots in floors
- Longer life and durability when compared to traditional and diamond floor pads
- A more sustainable solution as they can be used with just water to clean and maintain floors
- Longer brush life means less waste in landfills
- Coating-free maintenance system - no need for chemicals for floor stripping or finishing
- Brush can be used with just water, providing a more sustainable solution
- Designed for daily use with water on a low-speed machine (175 to 600 min⁻¹)
It is conformable, allowing the flexible bristles to reach low spots on the floor while minimising debris clogging. No assembly or pre-soaking is required and it is easy to install on standard, low-speed equipment.