Screw vials, Thermo Scientific™ Chromacol™
HYPE1.1-STVGN
- Lock storlek:8-425
- Diam×H:12×32 mm
- Kapacitet:1,10 ml
- Färg:Klar
- Beskrivning:Screw vial
- Förp.:500
Thermo Scientific™ Chromacol™ sample storage screw thread vials are manufactured from superior quality 33 expansion borosilicate glass, clear (Type 1, Class A).
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample.