SPE Plate, 96-well, NBE TRACE-N20
Catalog # TCAT611-0051NBE
Leverantör: Tecan
Specifikationer
- Beskrivning:Plate NBE TRACE-N20, 1ml, 20 µm
- Phase:TRACE-N20
- Fyllning:5 mg
- Förp.:1
Specifikationer
Om denna produkt
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
- Reversed-phase
- Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.