BD Vacutainer® Eclipse™ Signal™ Blood Collection Needle
Catalog # BDAM368835
Supplier: BD Medical
Specifications
- Mätare:G 21
- Beskrivning:BD Vacutainer® Eclipse™ Signal™ blood collection needle with integrated holder
- Längd:25 mm
- Förp.:50
Specifications
About this item
The BD Vacutainer® Eclipse™ Signal™ blood collection needles offers a clear dedicated window for in-vein confirmation with an extra thin-wall cannula for a fast blood collection experience.
The BD Eclipse™ Signal™ with the integrated holder offers an ergonomic shape and an integrated safety shield to support end user experience. Altogether, this can provide greater confidence during venous blood collection with ease of handling and increase healthcare worker safety.
Packaging: 50 per box, 400 per case