Centrifuge Tube Slicer
BECM347960
- Beskrivning:CentriTube slicer
- Förp.:1
This CentriTube slicer is used for slicing polypropylene, polycarbonate, and cellulose propionate centrifuge tubes with diameters of 5 to 13 mm.
This CentriTube slicer is used for slicing polypropylene, polycarbonate, and cellulose propionate centrifuge tubes with diameters of 5 to 13 mm.