High Visibility Fleece Jackets, F250
Leverantör: Portwest
Certifikat
Om denna produkt
The Hi-Vis essential fleece offers great wearer warmth and comfort. Features include two secure side pockets with zips and an elasticated draw cord waist for a more comfortable fit.
- Anti-pill durable fleece fabric
- Reflective tape for increased visibility
- Elastic waist with toggle for a secure and comfortable fit
- Elasticated cuffs for a secure fit
- Contrast navy collar and zip for added style
- 40+ UPF rated fabric to block 98% of UV rays
- Unlined
The high quality anti-pill finish maintains the look of the garment for longer.
Certifications: Certified to EN ISO 20471 Class 3, AS/NZS 4602.1 Class D/N.
Specifikationer
- Catalog No:
- PRTWF250YERXXXL
- PRTWF250YERL
- PRTWF250YERM
- PRTWF250YERXL
- PRTWF250YERS
- PRTWF250YERXXL
- PRTWF250YERXS
- Modell:
- F250
- F250
- F250
- F250
- F250
- F250
- F250
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Cold, HiVis
- Cold, HiVis
- Cold, HiVis
- Cold, HiVis
- Cold, HiVis
- Cold, HiVis
- Cold, HiVis
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester