ContactUsLinkComponent
Cerex® RCH PPBA 96 Well Plate
Cerex® RCH PPBA 96 Well Plate
Catalog # TCAT30162759
Supplier:  Tecan
Cerex® RCH PPBA 96 Well Plate
Catalog # TCAT30162759
Supplier:  Tecan
Specifications
  • Ant. brunnar:
    96
  • Beskrivning:
    Plate RCH Cerex® PPBA 1 ml
  • Phase:
    RCH
  • Fyllning:
    50 mg
  • Förp.:
    1
Specifications
About this item
A special phenyl boronic acid chemistry on polymer base resin for better capacity and solvent use.
  • Cerex
  • Reversed-phased
  • Liquid handler compatible
PBA utilises an unique covalent interaction ideal for catecholamine analysis.
Packaging: 1 piece per box