Cerex® ETGS 96 Well Plate, RCH
Katalog # TCAT642-0501RCH
Leverantör: Tecan
Specifikationer
- Beskrivning:Plate RCH Cerex ETGS, 1 ml, 10 µm
- Phase:ETGS
- Fyllning:50 mg
- Förp.:1
Specifikationer
Om denna produkt
A 10 µm micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
- Cerex
- Ion-exchange
- Liquid handler compatible
Formulated for ETS/ETGS Testings.
Packaging: 1 piece per box