ISO Humidity Calibration Certificate
TEST0520.0166
: TESTO
- Beskrivning:ISO humidity calibration certificate - with 3 measuring points
- Förp.:1
This ISO calibration certificate certifies that your measuring instrument or probe is calibrated. Calibration is carried out according to the ISO standard.
- Three calibration points: 11,3/50/75,3% RH at +25 °C
- According to the ISO standard
- Product/housing material: Paper
- Product colour: White