Om denna produkt
Utilizing a unique formulation, AlphaTec® 58-201 gloves offer a versatile protective solution that can be applied to a wide range of applications and environments.
- AnsellGrip Technology for enhanced grip in wet and oily conditions
- Designed to protect at temperatures from −40 to +250 °C (482 °F)
- Acrylic liner keeps hands warm in cold conditions and protected in contact heat applications
- Nitrile chemical barrier provides protection against a wide range of chemicals
AlphaTec® 58-201 gloves are recommended for use in all food processing applications that have food contact, cold storage, seafood processing, chemical handling, chemical processing and preparation, janitorial use, cleaning, and maintenance applications.
Enhanced grip: AlphaTec® 58-201 chemical gloves feature Ansell Grip™ Technology, which ensures a stronger and more secure hold in wet and oily environments. Heightened defenses: These cold and heat-resistant gloves are also designed to keep hands safe in temperatures ranging from−40 to +250 °C while providing chemical protection.
Elevated comfort: Thanks to their acrylic liner, AlphaTec® 58-201 green safety gloves deliver warmth in cold environments and delay heat transfer in contact heat applications. Added protection: These nitrile-coated gloves ensure superior chemical resistance, making them ideal for diverse applications and environments.
Specifikationer
- Catalog No:
- ANSE58-201/10
- ANSE58-201/8
- ANSE58-201/9
- Modell:
- 58-201
- 58-201
- 58-201
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Fodermaterial:
- Akryl
- Akryl
- Akryl
- Kragmodell:
- Kraghandske
- Kraghandske
- Kraghandske
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Antistatisk:
- +
- +
- +
- Godkända för livsmedelskontakt:
- +
- +
- +