Troston® Flame Retardant Antistatic Fleece Lining with Sleeves
Leverantör: SIOEN
Certifikat
Om denna produkt
Nothing is more comfortable and warm than a fleece lining.
- Round neck
- One patched chest pocket
- Round sleeves
- Knitted wind cuffs at end of sleeves
- Not to be worn separately
- Back length 750 mm
- Fixed flame retardant modacryl lining
- One inside pocket
In addition, the Troston is also flame retardant and even antistatic and Interchangeable Lining System (I.L.S.) can it easily be zipped into multiple jackets.
Packaging: Five pieces per box.
Specifikationer
- Catalog No:
- SIOE7870A2TF6/XL
- SIOE7870A2TF6/S
- SIOE7870A2TF6/M
- SIOE7870A2TF6/L
- SIOE7870A2TF6/3XL
- SIOE7870A2TF6/2XL
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST
- 54% Modacryl, 34,2% FR cotton, 10% Nylon, 1,8% AST