Smart® FFP Masks with Comfortable Nose Seal
Catalog # MOLD238501
Supplier: Moldex
Specifications
- Beskrivning:FFP1 NR D with ventex® valve
- Förp.:20
Specifications
About this item
Smart FFP masks are designed with a soft foam nose cushion for a comfortable fit. A soft, skin-friendly inner fleece layer and a stretchy woven strap provide additional comfort for the wearer.
- Reliable protection against particles
- Soft nose cushion for enhanced wearing comfort
- ActivForm® 3D design for a comfortable and secure fit
The flexible ActivForm® 3D design adapts automatically to the shape of the wearers face and their facial movements. As a result, our Smart masks are easy to wear and sit securely and comfortably on the skin at all times, without the need for a nose clip. Our DuraMesh® structure preserves the shape of the mask under even the most testing conditions.
Smart FFP masks offer reliable protection against particles.
Shelf life from date of production: 5 years.