Microflex® 92-134 Disposable Nitrile Examination Gloves
Supplier: Ansell
Certificates
About this item
Comfortable gloves offering tactility and protection across numerous applications.
- Soft nitrile formulation: Protective gloves offering superior comfort
- Lightweight construction: Nitrile gloves with enhanced tactility
- Textured fingertips: For secure handling of tools and materials
Ansell MICROFLEX® 92-134 versatility disposable safety gloves are made of a soft nitrile formulation, which provides a highly comfortable fit and feel while protecting hands. These nitrile disposable gloves lightweight construction provides augmented tactile sensitivity and fine motor control.
These MICROFLEX® gloves are textured on the fingertips to ensure a stronger grip for the safer handling of tools and materials. Designed for a broad range of applications, these gloves reinforce hand protection across a wide range of industries.
Certifications: ASTM D6319. CE 0493. EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 1-4. FDA21 CFR 177-2600, Food Contact. ISO 13485, ISO 9001. US QSR/FDA 510(k) Medical Examination Grade.
Packaging: 100 gloves per dispenser, 10 dispensers per case, 1000 gloves per case.
Specifications
- Catalog No:
- ANSE92-134/M
- ANSE92-134/S
- ANSE92-134/XL
- ANSE92-134/XS
- ANSE92-134/XXL
- ANSE92-134/L
- Modell:
- Microflex® 92-134
- Microflex® 92-134
- Microflex® 92-134
- Microflex® 92-134
- Microflex® 92-134
- Microflex® 92-134
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Material:
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Yta:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Manchettlängd:
- Standard
- Standard
- Standard
- Standard
- Standard
- Standard
- Finger, tjocklek:
- 4.3 mil
- 4.3 mil
- 4.3 mil
- 4.3 mil
- 4.3 mil
- 4.3 mil
- Handflata, tjocklek:
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- FDA-reglerad:
- +
- +
- +
- +
- +
- +
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +
- +
- ASTM nr.:
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319